Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Influenza B M Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111968
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111968 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111968 Supplier Invitrogen™ Supplier No. PA5111968
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Influenza B M Polyclonal Antibody for Western Blot, ICC/IF

The Influenza B matrix protein (M1) is a fundamental structural protein that plays a pivotal role in the virus lifecycle, providing shape and rigidity to the viral particle. The M1 protein lines the inner surface of the viral lipid envelope and interacts with both the ribonucleoprotein (RNP) complex and the cytoplasmic tails of the viral glycoproteins, hemagglutinin (HA), and neuraminidase (NA). This interaction is crucial for virus assembly, budding, and release from infected cells. The M1 protein also facilitates the transport of the viral RNP from the nucleus to the cytoplasm during virus replication. The highly conserved nature of the M1 protein makes it a potential target for antiviral strategies and vaccine development. Recent research has focused on understanding the precise mechanisms of M1's interaction with viral and host components to identify novel therapeutic targets.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Influenza B M
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.01% thimerosal; pH 7
Gene Alias Influenza B
Host Species Rabbit
Immunogen Recombinant fragment corresponding to Internal region of Influenza B virus M (B/Taiwan/753/2005). The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.