Learn More
Invitrogen™ Influenza B NA Monoclonal Antibody (B19)

Description
MA1-10825 detects Influenza B Neuraminidase. MA1-10825 has been successfully used in ELISA and inhibition applications. The MA1-10825 immunogen is Influenza B HK/73.
Specifications
Specifications
| Antigen | Influenza B NA |
| Applications | ELISA, Inhibition Assays |
| Classification | Monoclonal |
| Clone | B19 |
| Concentration | 0.9 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene Alias | Influenza NA |
| Host Species | Mouse |
| Immunogen | Influenza B HK/73. |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.