Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Influenza B nucleoprotein Antibody (016), DyLight 550, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP312755R
Description
Influenza B nucleoprotein Monoclonal antibody specifically detects Influenza B nucleoprotein in Virus samples. It is validated for Western Blot, ELISASpecifications
Influenza B nucleoprotein | |
Monoclonal | |
DyLight 550 | |
Influenza B Virus NP, NP, Nucleoprotein | |
This antibody was obtained from a rabbit immunized with purified, recombinant Influenza B (Accession#: ACF54251.1; Met1-Tyr560). | |
100 μg | |
Primary | |
Store at 4°C in the dark. | |
IgG |
Western Blot, ELISA | |
016 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Virus | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction