Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Influenza B nucleoprotein Antibody (3E9/B2) - (B/Lee/40) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$390.00
Specifications
| Antigen | Influenza B Virus Nucleoprotein |
|---|---|
| Clone | 3E9/B2 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000 |
| Applications | Western Blot, ELISA, Immunocytochemistry, Immunofluorescence |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP22351301
![]() |
Novus Biologicals
NBP2235130.1ML |
0.1 mL |
Each of 1 for $390.00
|
|
|||||
Description
Influenza B Virus Nucleoprotein Monoclonal specifically detects Influenza B Virus Nucleoprotein in Virus samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
| Influenza B Virus Nucleoprotein | |
| 1 mg/mL | |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Influenza B Virus NP, NP, Nucleoprotein | |
| IgG2a κ | |
| Protein A or G purified | |
| Clone 3E9/B2 reacts with influenza B virus nucleoprotein as demonstrated by immunoblotting after SDS-PAGE. Clone 3E9/B2 does not cross-react with influenza A virus. |
| 3E9/B2 | |
| Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000 | |
| Monoclonal | |
| Purified | |
| Virus | |
| Unpurified influenza B virus (B/Lee/40) for primary intranasal immunization, boosted intravenously with purified influenza B virus disrupted with Triton X-100 for 40 min at 37C. | |
| Primary | |
| Store at 4C. Do not freeze. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title