Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Influenza B Virions Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA17225
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA17225 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA17225 Supplier Invitrogen™ Supplier No. PA17225
Add to Cart
Add to Cart

Goat Polyclonal Antibody

PA1-7225 detects the influenza B virus. This antibody does not cross-react with Influenza A, Parainfluenza 1-3, RSV or Adenovirus. PA1-7225 has been successfully used in immunofluorescence procedures. The PA1-7225 immunogen is Yamagata strain of influenza B. The potential applications for this product are ELISA, IF, WB and IHC. The appropriate dilutions should be determined by the user.

Influenza A and B are the two types of influenza viruses that cause epidemic human disease. Influenza B viruses currently circulating are divided into two antigenically and genetically distinct groups: the Victoria and Yamagata lineages. Influenza virus B is a genus in the virus family Orthomyxoviridae. A limited host range means that Influenza virus B pandemics are rare. The nucleoprotein protects the negative strand viral RNA from nucleases by encapsidating it. Encapsidated genomic RNA is termed the ribonucleoprotein (RNP) and serves as template for transcription and replication.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Influenza B Virions
Applications Immunocytochemistry
Classification Polyclonal
Concentration 4-5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.2
Gene Alias Influenza Virions
Host Species Goat
Immunogen Yamagata strain of Influenza B.
Purification Method IgG Fraction
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.