Learn More
Invitrogen™ Influenza B Virions Polyclonal Antibody, Biotin

Description
The PA1-73038 antibody reacts with Influenza B Virions. PA1-73038 has been successfully used in Immunohistochemical, ELISA, Immunofluorescence and Western blot applications. The PA1-73038 antibody was raised against the Yamagata strain of Influenza B.
Specifications
Specifications
| Antigen | Influenza B Virions |
| Applications | ELISA, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 4-5 mg/mL |
| Conjugate | Biotin |
| Formulation | PBS with 0.1% sodium azide; pH 7.2 |
| Gene Alias | Influenza Virions |
| Host Species | Goat |
| Immunogen | Yamagata strain of Influenza B. |
| Purification Method | IgG fraction |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.