Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Influenza B Hemagglutinin 2 Antibody (InB18), Alexa Fluor™ 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10073163AF488
Description
Influenza B Hemagglutinin 2 Monoclonal antibody specifically detects Influenza B Hemagglutinin 2 in Virus samples. It is validated for Western Blot, ELISA.Specifications
Influenza B Hemagglutinin 2 | |
Monoclonal | |
Alexa Fluor 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with Influenza B virus. | |
0.2 mL | |
Primary | |
Virus | |
Purified |
Western Blot, ELISA | |
InB18 | |
Western Blot, ELISA | |
Mouse | |
Protein A purified | |
RUO | |
Influenza B haemagglutinin 2 (HA2) | |
Store at 4C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction