Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Influenza H18N11 HA1 (A/flat-faced bat/Peru/033/2010) Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA581755
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Influenza H18N11 (A/flat-faced bat/Peru/033/2010) Hemagglutinin/HA1.

Specifications
Influenza H18N11 HA1 (A/flat-faced bat/Peru/033/2010) | |
Polyclonal | |
Unconjugated | |
flu hemagglutinin; influenza hemagglutinin | |
Recombinant Influenza H18N11 (A/flat-faced bat/Peru/033/2010) Hemagglutinin/HA1 Protein. | |
100 μL | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Western Blot | |
1 mg/mL | |
PBS with no preservative | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
Virus | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction