Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Influenza H3N2 HA1 (A/reassortant/IVR-155(Victoria/210/2009 x Puerto Rico/8/1934)) Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA581735
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Influenza H3N2 (A/reassortant/IVR-155(Victoria/210/2009 x Puerto Rico/8/1934)) Hemagglutinin/HA1.

Specifications
Influenza H3N2 HA1 (A/reassortant/IVR-155(Victoria/210/2009 x Puerto Rico/8/1934)) | |
Polyclonal | |
Unconjugated | |
flu hemagglutinin; influenza hemagglutinin | |
Recombinant Influenza H3N2 (A/reassortant/IVR-155(Victoria/210/2009 x Puerto Rico/8/1934)) Hemagglutinin/HA1 Protein. | |
100 μL | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with no preservative | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
Virus | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction