Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Influenza H5N1 HA Recombinant Human Monoclonal Antibody (H011)
GREENER_CHOICE

Catalog No. PIMA529949
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA529949 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA529949 Supplier Invitrogen™ Supplier No. MA529949
Only null left
Add to Cart
Add to Cart

Human Recombinant Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant antibodies are produced using specific genes that code for the desired antibodies. These genes are cloned into an expression vector and expressed in vitro. The advantages of recombinant antibodies include: better specificity, animal origin-free formulation, and more lot-to-lot consistency. This antibody has specificity for H5N1 (A/Hubei/1/2010) HA H5N1 (A/Xinjiang/1/2006) HA1 H5N1 (A/Xinjiang/1/2006) HA H5N1 (A/bar-headed goose/Qinghai/1A/2005) HA H5N1 (A/Anhui/1/2005) HA H5N1 (A/Cambodia/S1211394/2008) HA H5N1 (A/Duck/Hong Kong/p46/97) HA H5N1 (A/Hong kong/213/2003) HA (28 Ser/Trp) H5N1 (A/chicken/India/NIV33487/06) HA H5N1 (A/Cambodia/R0405050/2007) HA H5N1 (A/whooper swan/Mongolia/244/2005) HA H5N1 (A/Egypt/N05056/2009) HA H5N1 (A/Common magpie/Hong Kong/2256/2006) HA H5N1 (A/duck/Hunan/795/2002) HA H5N1 (A/Egy.

Influenza A virus is a major public health threat, killing more than 30,000 people per year in the USA. In early 2009, a novel swine-origin influenza A (H1N1) virus was identified in specimens obtained from patients in Mexico and the United States. The virus spread quickly around the world and on June 11, 2009, the World Health Organization declared it a pandemic. Influenza A virus has one of sixteen possible Hemagglutinin (HA) surface proteins and one of nine possible Neuraminidase (NA) surface proteins. The Hemagglutinin protein facilitates viral attachment while Neuraminidase is involved in viral release. These proteins also elicit immune responses that prevent infection or independently reduce viral replication. The genetic make-up of this swine flu virus is unlike any other: it is an H1N1 strain that combines a triple assortment first identified in 1998 including human, swine, and avian influenza with two new pig H3N2 virus genes from Eurasia, themselves of recent human origin. The distinct antigenic properties of the new swine influenza virus compared with seasonal influenza A (H1N1) virus suggest that human immunity against new swine influenza virus is limited, although the age distribution of reported cases suggests some degree of protection in older age groups.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Influenza H5N1 HA
Applications Inhibition Assays, Neutralization
Classification Recombinant Monoclonal
Clone H011
Concentration 4.09 mg/mL
Conjugate Unconjugated
Formulation Arginine, Histidine with 120mM NaCl, 0.02% Tween 80 and no preservative; pH 6
Gene Alias flu hemagglutinin; influenza hemagglutinin
Host Species Human
Immunogen Influenza H5N1 Hemagglutinin/HA protein.
Purification Method Size-exclusion - Membrane filtration
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.