Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
InfluenzaBYamagata Goat anti-Virus, Polyclonal, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NBP266600
Description
Influenza B Yamagata Polyclonal specifically detects Influenza B Yamagata in Virus samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.Specifications
InfluenzaBYamagata | |
Polyclonal | |
Western Blot 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500 | |
Influenza virus, Influenza-B | |
Influenza B virions, Yamagata strain. | |
1 mg | |
Virus | |
Purified |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Western Blot | |
Unconjugated | |
PBS (pH 7.5) before the addition of 40% glycerol with 0.1% Sodium Azide | |
Goat | |
>95% | |
Primary | |
Store at 4°C after rehydration. For extended storage after rehydration, add an equal volume of glycerol and store at -20°C. Avoid repeated freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction