Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ING1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP131006
Description
ING1 Polyclonal specifically detects ING1 in Human samples. It is validated for Western Blot.Specifications
ING1 | |
Polyclonal | |
Unconjugated | |
PBS (pH 7.0) and 40% Glycerol with 0.01% Thimerosal | |
ING1 | |
Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human ING1. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
Primary | |
Tested in human. Predicted cross-reactivity based on sequence identity: Rhesus Monkey (100%). Not yet tested in any other species. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
1.0mg/mL | |
Western Blot 1:500-1:3000 | |
growth inhibitor ING1, growth inhibitory protein ING1, inhibitor of growth family, member 1, inhibitor of growth protein 1, p24ING1c, p33, p33ING1, p33ING1b, p47, p47ING1a, tumor suppressor ING1 | |
Rabbit | |
47 kDa | |
100 μL | |
Apoptosis, Cancer, Chromatin Research, p53 Pathway, Transcription Factors and Regulators, Tumor Suppressors | |
3621 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction