Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ING1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA596854
Description
Positive Samples: SKOV3, HT-29, NIH/3T3, Mouse testis, Mouse brain, Mouse spleen, Mouse lung, Rat brain, Rat liver; Cellular Location: Nucleus
This gene encodes a tumor suppressor protein that can induce cell growth arrest and apoptosis. The encoded protein is a nuclear protein that physically interacts with the tumor suppressor protein TP53 and is a component of the p53 signaling pathway. Reduced expression and rearrangement of this gene have been detected in various cancers. Multiple alternatively spliced transcript variants encoding distinct isoforms have been reported.
Specifications
ING1 | |
Polyclonal | |
Unconjugated | |
ING1 | |
2610028J21Rik, AA407184, AI875420, growth inhibitor ING1, growth inhibitory protein ING1, ING1, inhibitor of growth family member 1, inhibitor of growth family, member 1, inhibitor of growth protein 1, mING1h, p24ING1c, p33, p33ING, p33ING1, p33ING1b, p33ING1c, p37Ing1b, p47, p47ING1a, tumor suppressor ING1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
26356, 306626, 3621 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
6.15 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide, pH 7.3 | |
Q9QXV3, Q9UK53 | |
ING1 | |
A synthetic peptide of human ING1. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction