Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ING2 Antibody (AT39E5), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP261159H
This item is not returnable.
View return policy
Description
ING2 Monoclonal antibody specifically detects ING2 in Human samples. It is validated for Western Blot, ELISASpecifications
ING2 | |
Monoclonal | |
HRP | |
ING1Linhibitor of growth family, member 1-like, ING1Lp, Inhibitor of growth 1-like protein, inhibitor of growth family, member 2, inhibitor of growth protein 2, p33ING2p32 | |
Recombinant human ING2 (1-280aa) purified from E. coli | |
0.1 mL | |
Apoptosis, DNA Repair | |
3622 | |
Store at 4°C in the dark. | |
IgG1b κ |
Western Blot, ELISA | |
AT39E5 | |
PBS | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction