Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ING2 Goat anti-Human, Polyclonal, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NB1001044100UG
Description
ING2 Polyclonal specifically detects ING2 in Human samples. It is validated for Western Blot, Peptide ELISA.Specifications
ING2 | |
Polyclonal | |
Unconjugated | |
Tris (pH 7.3), 0.5% BSA | |
ING1Linhibitor of growth family, member 1-like, ING1Lp, Inhibitor of growth 1-like protein, inhibitor of growth family, member 2, inhibitor of growth protein 2, p33ING2p32 | |
Peptide with sequence C-DKSTEKTKKDRRSR corresponding to C-Terminus according to NP_001555.1. | |
100 μg | |
Apoptosis, DNA Repair | |
3622 | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot, ELISA | |
0.5 mg/ml | |
Western Blot 1 - 3 ug/mL, Peptide ELISA Detection limit 1:1000 | |
NP_001555 | |
Goat | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction