Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ING2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554434
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554434 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554434 Supplier Invitrogen™ Supplier No. PA554434
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MLGQQQQQLY SSAALLTGER SRLLTCYVQD YLECVESLPH DMQRNVSVLR ELDNKYQETL KEIDDVYEKY KKEDDLNQKK RLQQLLQRAL Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 97%.

This gene is a member of the inhibitor of growth (ING) family. Members of the ING family associate with and modulate the activity of histone acetyltransferase (HAT) and histone deacetylase (HDAC) complexes and function in DNA repair and apoptosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ING2
Applications ChIP Assay, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Ing2
Gene Accession No. Q9H160
Gene Alias 2810011M06Rik; ING1L; ING1Lp; ING2; Ing2b; Inhibitor of growth 1-like protein; inhibitor of growth family member 2; inhibitor of growth family, member 2; inhibitor of growth protein 2; p32; p33ING2
Gene Symbols Ing2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ING2. Recombinant protein control fragment (Product #RP-91875).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3622
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.