Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ING2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114750
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114750 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114750 Supplier Invitrogen™ Supplier No. PA5114750
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ING2.

This gene is a member of the inhibitor of growth (ING) family. Members of the ING family associate with and modulate the activity of histone acetyltransferase (HAT) and histone deacetylase (HDAC) complexes and function in DNA repair and apoptosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ING2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene Ing2
Gene Accession No. Q9ESK4, Q9H160
Gene Alias 2810011M06Rik; ING1L; ING1Lp; ING2; Ing2b; Inhibitor of growth 1-like protein; inhibitor of growth family member 2; inhibitor of growth family, member 2; inhibitor of growth protein 2; p32; p33ING2
Gene Symbols Ing2
Host Species Rabbit
Immunogen A synthesized peptide derived from human ING2(Accession Q9H160), corresponding to amino acid residues Y247-R280.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3622, 69260
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.