Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ING3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566934
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566934 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566934 Supplier Invitrogen™ Supplier No. PA566934
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MLYLEDYLEMI EQLPMDLRDR FTEMREMDLQ VQNAMDQLEQ RVSEFFMNAK KNKPEWREEQ MASIKKDYYK ALEDADEKVQ LANQI Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.

The protein encoded by this gene is similar to ING1, a tumor suppressor protein that can interact with TP53, inhibit cell growth, and induce apoptosis. This protein contains a PHD-finger, which is a common motif in proteins involved in chromatin remodeling. This gene can activate p53 trans-activated promoters, including promoters of p21/waf1 and bax. Overexpression of this gene has been shown to inhibit cell growth and induce apoptosis. Allelic loss and reduced expression of this gene were detected in head and neck cancers. Two alternatively spliced transcript variants encoding different isoforms have been observed.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ING3
Applications Immunocytochemistry
Classification Polyclonal
Concentration 1.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ING3
Gene Accession No. Q9NXR8
Gene Alias 1300013A07Rik; Eaf 4; Eaf4; FLJ20089; HSPC301; ING 2; ING 3; ING2; ING3; ing3 protein; inhibitor of growth family member 3; inhibitor of growth family, member 3; inhibitor of growth protein 3; inhibitor of growth protein 3-like protein; MEAF4; p47 p47 ING3; p47 regulator protein; p47ING3; p47ING3 protein; zgc:56327
Gene Symbols ING3
Host Species Rabbit
Immunogen Recombinant Human ING3. Recombinant protein control fragment (Product #RP-105430).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54556
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.