Learn More
ING3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 109051AP150UL
Description
This antibody is specifically against p47ING3.
ING3 belongs to a family of proteins containing the plant homeodomain (PHD) finger, which includes transcription factors and proteins that regulate chromatin structure. ING3 is a component of the NuA4 histone acetyltransferase (HAT) complex and can activate p53 trans-activated promoters, including promoters of p21/waf1 and bax. Overexpression of ING3 has been shown to inhibit cell growth and induce apoptosis. Allelic loss and reduced expression of the ING3 gene were detected in head and neck cancers.Specifications
| ING3 | |
| Polyclonal | |
| Unconjugated | |
| ING3 | |
| Eaf4, ING2, ING3, Inhibitor of growth protein 3, p47ING3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 54556, 71777 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8VEK6, Q9NXR8 | |
| ING3 | |
| ING3 Fusion Protein Ag1357 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.