Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ING4 Antibody, Alexa Fluor™ 750, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NB1001682AF750
Description
ING4 Polyclonal specifically detects ING4 in Human samples. It is validated for Western Blot, ELISA.Specifications
ING4 | |
Polyclonal | |
Western Blot, ELISA | |
candidate tumor suppressor p33 ING1 homolog, inhibitor of growth family, member 4, inhibitor of growth protein 4, MGC12557, my036, p29ING4brain my036 protein | |
This affinity purified ING4 Antibody was prepared from whole goat serum produced by repeated immunizations with a synthetic peptide corresponding to an internal region near aa 150-175 of Human ING4 protein (Inhibitor of growth family, member 4). This sequence only shows homology to isoform 1 for ING4. (Uniprot: Q9UNL4) | |
0.1 mL | |
Chromatin Research, Tumor Suppressors | |
51147 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, ELISA | |
Alexa Fluor 750 | |
50mM Sodium Borate | |
Goat | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction