Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ING5 Antibody (119), mFluor Violet 610 SE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290261MFV610
Description
ING5 Monoclonal antibody specifically detects ING5 in Human samples. It is validated for Western Blot, ELISA, ImmunoprecipitationSpecifications
ING5 | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, ELISA, Immunoprecipitation | |
119 | |
Western Blot, ELISA, Immunoprecipitation | |
inhibitor of growth family, member 5, inhibitor of growth protein 5, p28ING5FLJ23842 | |
Rabbit immunized with purified, recombinant Human ING5 (Uniprot#: Q8WYH8-2, Glu36-Gln84) | |
0.1 mL | |
Cancer, Tumor Suppressors | |
84289 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction