Learn More
Invitrogen™ INHA Polyclonal Antibody

Description
INHA Polyclonal Antibody for Flow, ICC/IF, IHC (P), Western Blot
Specifications
Specifications
| Antigen | INHA |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1.8 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | INHA |
| Gene Accession No. | P05111, Q04997 |
| Gene Alias | A inhibin subunit; A-inhibin subunit; AW555078; IHA; Inha; inhibin; inhibin alpha; inhibin alpha chain; inhibin alpha subunit |
| Gene Symbols | INHA |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.