Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

INHAT-1 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 1 publication

$181.90 - $402.00

Specifications

Antigen INHAT-1
Concentration 0.5 mg/mL
Dilution Western Blot 1-2 ug/ml
Applications Western Blot
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB10056353T
SDP
View Documents
Novus Biologicals
NB10056353SS
0.025 mg
Each for $181.90
Only null left
Add to Cart
 
NB10056353
SDP
View Documents
Novus Biologicals
NB10056353
0.1 mg
Each for $402.00
Only null left
Add to Cart
 
Description

Description

INHAT-1 Polyclonal specifically detects INHAT-1 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications

Specifications

INHAT-1
Western Blot 1-2 ug/ml
Polyclonal
Purified
RUO
0.2 ml PBS and 0.1 mg/ml BSA with 0.02% Sodium Azide
This antibody was raised against synthetic peptides of amino acids 66-81 and 135-151 of human INHAT-1. Sequence homology is 100% in mouse and rat.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
0.5 mg/mL
Western Blot
Unconjugated
Rabbit
Human, Mouse, Rat
TAF-1a, TAF-1b
IgG
Protein G purified
An approx. 37 kDa band is observed.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.