Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ INHBB Monoclonal Antibody (C11), Invitrogen™
GREENER_CHOICE

Catalog No. PIMA543691
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA543691 100 μL
PIMA543692 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIMA543691 Supplier Invitrogen™ Supplier No. MA543691
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

The inhibin beta B subunit joins the alpha subunit to form a pituitary FSH secretion inhibitor. Inhibin has been shown to regulate gonadal stromal cell proliferation negatively and to have tumour-suppressor activity. In addition, serum levels of inhibin have been shown to reflect the size of granulosa-cell tumors and can therefore be used as a marker for primary as well as recurrent disease. Because expression in gonadal and various extragonadal tissues may vary severalfold in a tissue-specific fashion, it is proposed that inhibin may be both a growth/differentiation factor and a hormone. Furthermore, the beta B subunit forms a homodimer, activin B, and also joins with the beta A subunit to form a heterodimer, activin AB, both of which stimulate FSH secretion.
TRUSTED_SUSTAINABILITY

Specifications

Antigen INHBB
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone C11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene INHBB
Gene Accession No. P09529
Gene Alias activin AB beta polypeptide; activin betaB; activin beta-B chain; INHBB; Inhibin beta B chain; inhibin beta B subunit; inhibin beta subunit B; inhibin beta-B; inhibin, beta B; Inhibin, beta-2
Gene Symbols INHBB
Host Species Mouse
Immunogen Recombinant INHbB (Gly293-Ala407) expressed in E. coli.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3625
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.