Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ INHBC Monoclonal Antibody (CL6916)
GREENER_CHOICE

Catalog No. PIMA531444
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531444 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531444 Supplier Invitrogen™ Supplier No. MA531444
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Immunogen sequence: TPRAGGQCPA CGGPTLELES QRELLLDLAK RSILDKLHLT QRPTLNRPVS RAALRTALQH LHGVPQGALL EDNREQECEI Highest antigen sequence indentity to the following orthologs: Mouse - 65%, Rat - 63%.

Inhibins and activins are involved in regulating a number of functions such as hypothalamic and pituitary hormone secretion, gonadal hormone secretion, germ cell development and maturation, erythroid differentiation, insulin secretion, nerve cell survival, embryonic axial development or bone growth depending on their subunit composition. Activins oppose the funtions of inhibins. Inhibins are predominantly expressed in liver, uterus and ovary tissue, but also in benign prostatic hyperplasia. Inhibin A, a dimer of alpha and beta-A, and inhibin B, a dimer of alpha and beta-B, have been shown to inhibit the secretion of follicle stimulating hormone. Inhibin beta-C forms a homodimer and its expression is predominant in adult liver and in benign prostatic hyperplasia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen INHBC
Applications Western Blot
Classification Monoclonal
Clone CL6916
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene INHBC
Gene Accession No. P55103
Gene Alias activin beta-C chain; activin/inhibin beta C; IHBC; Inhbc; inhibin beta C; inhibin beta C chain; inhibin beta C subunit; inhibin beta-C; inhibin, beta C
Gene Symbols INHBC
Host Species Mouse
Immunogen Recombinant protein fragment of Human INHBC (Product #RP-108706)
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3626
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.