Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ INHBC Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA548063
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA548063 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA548063 Supplier Invitrogen™ Supplier No. PA548063
Add to Cart
Add to Cart

Goat Polyclonal Antibody

In direct ELISA, approximately 5% cross-reactivity with recombinant human (rh) Activin A and rhInhibin A is observed and no cross-reactivity with rhInhibin B is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

Inhibins and activins are involved in regulating a number of functions such as hypothalamic and pituitary hormone secretion, gonadal hormone secretion, germ cell development and maturation, erythroid differentiation, insulin secretion, nerve cell survival, embryonic axial development or bone growth depending on their subunit composition. Activins oppose the funtions of inhibins. Inhibins are predominantly expressed in liver, uterus and ovary tissue, but also in benign prostatic hyperplasia. Inhibin A, a dimer of alpha and beta-A, and inhibin B, a dimer of alpha and beta-B, have been shown to inhibit the secretion of follicle stimulating hormone. Inhibin beta-C forms a homodimer and its expression is predominant in adult liver and in benign prostatic hyperplasia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen INHBC
Applications Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene INHBC
Gene Accession No. P55104
Gene Alias activin beta-C chain; activin/inhibin beta C; IHBC; Inhbc; inhibin beta C; inhibin beta C chain; inhibin beta C subunit; inhibin beta-C; inhibin, beta C
Gene Symbols INHBC
Host Species Goat
Immunogen E. coli-derived recombinant mouse Activin C peptide (aa 236-352).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16325
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.