Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ INHBE Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553503
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553503 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553503 Supplier Invitrogen™ Supplier No. PA553503
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RPLEGNSTVT GQPRRLLDTA GHQQPFLELK IRANEPGAGR ARRRTPTCEP ATPLCCRRDH YVDFQELGWR DWILQPEGYQ LNYCSGQCPP HLAG Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 88%.

Inhibins and activins inhibit and activate, respectively, the secretion of follitropin by the pituitary gland. Inhibins/activins are involved in regulating a number of diverse functions such as hypothalamic and pituitary hormone secretion, gonadal hormone secretion, germ cell development and maturation, erythroid differentiation, insulin secretion, nerve cell survival, embryonic axial development or bone growth, depending on their subunit composition. Inhibins appear to oppose the functions of activins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen INHBE
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene INHBE
Gene Accession No. P58166
Gene Alias activin beta E; activin betaE; activin beta-E chain; INHBE; inhibin beta E; inhibin beta E chain; inhibin beta E subunit; inhibin, beta E
Gene Symbols INHBE
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human INHBE. Recombinant protein control fragment (Product #RP-92420).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83729
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.