Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Inhibin alpha Antibody (INHA/4266), Alexa Fluor™ 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308780AF488
Description
Inhibin alpha Monoclonal antibody specifically detects Inhibin alpha in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
Inhibin alpha | |
Monoclonal | |
Alexa Fluor 488 | |
A-inhibin subunit, INHA, inhibin alpha chain, inhibin, alpha | |
A synthetic peptide (around aa73-96) of alpha-subunit of human Inhibin alpha protein (exact sequence is proprietary) (Uniprot: P05111 ) | |
100 μg | |
Apoptosis, Cancer | |
3623 | |
Store at 4°C in the dark. | |
IgG2a |
Immunohistochemistry (Paraffin) | |
INHA/4266 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction