Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Inhibin alpha Antibody (OTI12F11), Alexa Fluor™ 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271039AF488
Description
Inhibin alpha Monoclonal specifically detects Inhibin alpha in Human samples. It is validated for Western Blot.Specifications
| Inhibin alpha | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| INHA | |
| Recombinant protein fragment corresponding to amino acids 62-232 of human Inhibin alpha (NP_002182) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot | |
| OTI12F11 | |
| Western Blot | |
| A-inhibin subunit, INHA, inhibin alpha chain, inhibin, alpha | |
| Mouse | |
| Affinity Purified | |
| Apoptosis, Cancer | |
| 3623.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction