Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Inhibin alpha Antibody (OTI12F11), Alexa Fluor™ 700, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271039AF700
Description
Inhibin alpha Monoclonal specifically detects Inhibin alpha in Human samples. It is validated for Western Blot.Specifications
Inhibin alpha | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
INHA | |
Recombinant protein fragment corresponding to amino acids 62-232 of human Inhibin alpha (NP_002182) produced in E.coli. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Western Blot | |
OTI12F11 | |
Western Blot | |
A-inhibin subunit, INHA, inhibin alpha chain, inhibin, alpha | |
Mouse | |
Affinity Purified | |
Apoptosis, Cancer | |
3623.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction