Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Inhibin alpha Antibody (PO12/8) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NBP261936 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP261936 0.1 mg
NB403017 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP261936 Supplier Novus Biologicals Supplier No. NBP261936
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Inhibin alpha Monoclonal specifically detects Inhibin alpha in Human samples. It is validated for ELISA, Immunohistochemistry.

Specifications

Antigen Inhibin alpha
Applications ELISA, Immunohistochemistry
Classification Monoclonal
Clone PO12/8
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution ELISA 90 ug/ml, Immunohistochemistry
Formulation PBS with 0.02% Sodium Azide
Gene Alias A-inhibin subunit, INHA, inhibin alpha chain, inhibin, alpha
Gene Symbols INHA
Host Species Mouse
Immunogen Synthetic peptide corresponding to epitope region aa73-96 of the alphaC region subunit of Inhibin alpha
Molecular Weight of Antigen 40 kDa
Purification Method Protein A purified
Quantity 0.1 mg
Research Discipline Apoptosis, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 3623
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.