Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Inhibin alpha Antibody (PO12/8), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP261936IR
Description
Inhibin alpha Monoclonal specifically detects Inhibin alpha in Human samples. It is validated for ELISA, Immunohistochemistry.Specifications
Inhibin alpha | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
INHA | |
Synthetic peptide corresponding to epitope region aa73-96 of the alphaC region subunit of Inhibin alpha | |
0.1 mL | |
Primary | |
Human | |
Purified |
ELISA, Immunohistochemistry | |
PO12/8 | |
ELISA, Immunohistochemistry | |
A-inhibin subunit, INHA, inhibin alpha chain, inhibin, alpha | |
Mouse | |
Protein A purified | |
Apoptosis, Cancer | |
3623 | |
Store at 4C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction