Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Inhibin alpha Antibody (R1), Janelia Fluor™ 549, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308781JF549
Description
Inhibin alpha Monoclonal antibody specifically detects Inhibin alpha in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
Inhibin alpha | |
Monoclonal | |
Janelia Fluor 549 | |
A-inhibin subunit, INHA, inhibin alpha chain, inhibin, alpha | |
A synthetic peptide corresponding to Human Inhibin alpha aa 233-264. Note that this peptide corresponds to the N-terminus of the mature region of the human inhibin alpha subunit. (Uniprot: P05111 ) | |
100 μg | |
Apoptosis, Cancer | |
3623 | |
Store at 4°C in the dark. | |
IgG2a κ |
Immunohistochemistry (Paraffin) | |
R1 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction