Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ INIP Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA554191
Description
Immunogen sequence: KEKRKLLMQN QSSTNHPGAS IALSRPSLNK DFRDHAEQQH IAAQQKAALQ HAHAHSSGYF ITQDSAFGNL Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 94%.

Specifications
INIP | |
Polyclonal | |
Unconjugated | |
INIP | |
1110054O05Rik; 2610312O17Rik; AA399876; C9orf80; HSPC043; HSPC291; hSSB-interacting protein 1; hSSBIP1; Inip; INTS3 and NABP interacting protein; INTS3- and NABP-interacting protein; minute INTS3/hSSB-associated element; MISE; RGD1307983; Sensor of single-strand DNA complex subunit C; sensor of ssDNA subunit C; Single-stranded DNA-binding protein-interacting protein 1; SOSS complex subunit C; SOSSC; SOSS-C; SSB-interacting protein 1; Ssbip1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
58493 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NRY2 | |
INIP | |
Recombinant protein corresponding to Human INIP. Recombinant protein control fragment (Product #RP-92999). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction