Learn More
Restek™ Injector Mounting and Parking Posts
SDP
Catalog No. 06611206 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Type:
Injector Mounting Post
Parking Post
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
06-611-206 Parking Post
06-611-195 Injector Mounting Post
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 06-611-206 Supplier Restek™ Supplier No. 22343
For Agilent 7673 & 7683 Autosamplers
  • Injector Mounting and Parking Posts
  • For Agilent 7683 Series Autosamplers, for use with 6850/6890 GCs
  • Performance equivalent to original manufacturer's parts
Specifications
Type Parking Post
Material Stainless Steel
For Use With (Application) Agilent 7673 & 7683 Autosamplers
Quantity 1
For Use With (Equipment) 5890, 6890, and 7890 GCs