Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ INK4b (p15), Mouse, Unlabeled, Clone: GF10 (10C7A6),

Catalog No. MABE1664MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABE1664MI 100 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE1664MI Supplier MilliporeSigma™ Supplier No. MABE1664
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cyclin-dependent kinase 4 inhibitor B (UniProt P42772; also known as MTS-2, Multiple tumor suppressor 2, p14-INK4b, p15-INK4b, p15INK4B) is encoded by the CDKN2B (also known as MTS2) gene (Gene ID 1030) in human. The INK and CIP/KIP families of cyclin dependent kinase inhibitors (CDKis) protect cells from oncogenic signals-initiated cellular transformation. INK4a and INK4b (encoded by the CDKN2A and CDKN2B gene, respectively) constitute the INK4 subfamily of CDKis. INK4a and INK4b are induced by oncogenic Ras and are involved in cell cycle G1 arrest and cellular senescence. Inactivation of CDKN2A and CDKN2B are found in a wide variety of human malignancies and murine tumor models.
TRUSTED_SUSTAINABILITY

Specifications

Antigen INK4b (p15)
Applications Western Blot
Classification Monoclonal
Clone GF10 (10C7A6)
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Mouse monoclonal IgG1κ hybridoma culture supernatant with 0.05% Sodium Azide.
Gene CDKN2B, MTS2
Gene Accession No. P42772
Host Species Mouse
Immunogen Recombinant full-length human INK4b (p15).
Purification Method Unpurified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Epigenetics & Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_004927
Test Specificity Clone GF10 (10C7A6), Mfr. No. MABE1664 and clone GF7 (7CIG4), Mfr. No. 05-430 correspond to two independent clones originated from the same mouse host. Clone GF10 (10C7A6) is recommended as an alternative to clone GF7 (7CIG4), however these two clones are expected to recognize different epitopes, exhibit different target-binding affinity and species cross-reactivity. Cross-reactivity of clone GF10 (10C7A6) toward spliced isoform 2 (p10; UniProt P42772-2) has not been confirmed.
Target Species Human
Content And Storage Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Form Culture Supernatant
Isotype IgG1 κ
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.