Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Inkjet Polyester Label and Overlaminate Kit

Catalog No. p-200009392 Shop All Brady Corporation Products
Click to view available options
Dimensions (LxW):
10 x 7 in.
3.35 x 2.17 in.
4.33 x 2.95 in.
6.1 x 4.13 in.
8.3 x 6.1 in.
For Use With (Equipment):
J2000 Printer
J4000, J5000 Printer

Take your inkjet printing further and label faster with these ideally-sized label and overlaminate kits.

  • Just peel, adhere and apply with these pre-sized labels and overlaminate
  • Inkjet-printable label is outfitted with a specially formulated topcoat that invigorates colors and label appearance
  • Label topcoat also protects against smudges, smears and splotches
  • Acrylic overlaminate provides up to 10 years of outdoor durability through improved UV and abrasion resistance
  • Overlaminate is oversized by a quarter-inch for easier application and complete coverage
  • An ink cartridge is required to print label material

Specifications

Autoclavable Not Autoclavable
Adhesive Type Permanent Acrylic
Coating None
Color White
Finish Matte
Format Die-cut
Material Polyester
Printable Sides Single-side Printable
Print Technology Ink Jet
Product Type Label
Sterility Non-sterile
For Use With (Application) Facility Identification, General Identification, Hazard Communication Identification, Lean and 5S Labeling, Lockout Tagout, Pipe and Valve Marking, Warehouse Marking
For Use With (Equipment) J2000 Printer
Includes 2-roll Kit
Min. Operating Temperature (Metric) -40°C
Quantity 340 Labels
Shape Rectangle
Size (Metric) 85.09 x 55.12 mm
Temperature Range (English) -40°F to +140°F
Temperature Range (Metric) -40°C to +60°C
Thickness (English) 0.0045 in.
Thickness (Metric) 0.1143 mm
Dimensions (LxW) 3.35 x 2.17 in.
Length (English) 3.35 in.
Length (Metric) 85.09 mm
Width (English) 2.17 in.
Width (Metric) 55.12 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.