Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Inkjet Vinyl Label

Catalog No. p-200009393 Shop All Brady Corporation Products
Click to view available options
Dimensions (LxW):
1 x 1 in.
1.5 x 1.5 in.
100 ft. x 1.125 in.
2 x 2 in.
For Use With (Equipment):
J2000 Printer
J4000 Printer
Format:
Continuous
Die-cut
Shape:
Rectangle
Round

Make an impact with this inkjet-printable vinyl label outfitted with a specially formulated topcoat that invigorates colors and label appearance.

  • Flexible material withstands harsh elements, including liquids, alcohol, chemicals, and indoor fluorescent and UV lighting
  • Topcoat also protects against smudges, smears and splotches
  • An ink cartridge is required to print this material

Specifications

Autoclavable Not Autoclavable
Adhesive Type Permanent Acrylic
Coating None
Color White
Finish Matte
Format Die-cut
Material Vinyl
Printable Sides Single-side Printable
Print Technology Ink Jet
Product Type Label
Sterility Non-sterile
For Use With (Application) Facility Identification, General Identification, Hazard Communication Identification, Lean and 5S Labeling, Lockout Tagout, Pipe and Valve Marking, Warehouse Marking
For Use With (Equipment) J2000 Printer
Min. Operating Temperature (Metric) -40°C
Quantity 1400 Labels
Shape Rectangle
Size (Metric) 25.4 x 25.4 mm
Temperature Range (Metric) -40°C to +60°C
Thickness (English) 0.0054 in.
Thickness (Metric) 0.137 mm
Dimensions (LxW) 1 x 1 in.
Length (English) 1 in.
Length (Metric) 25.4 mm
Width (English) 1 in.
Width (Metric) 25.4 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.