Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
METTLER TOLEDO™ InLab™ Pure pH Electrodes
Click to view available options
Connector Types:
Multi-Pin
S7
Description
- For use with METTLER TOLEDO SevenExcellence and SevenCompact meters as well as other brands. Connection cables may be required.
- Intelligent Sensor Management (ISM) electrodes capture and save calibration data, which is then automatically recognized by the meter once connected.
- Capture electrode calibration and results with LabX software, designed for advanced data management and 21CFR part 11 regulatory compliance.
- Compatible with InMotion Autosampler to automate measurement and cleaning of up to 303 samples.
Featured Applications
- Pure water
- Ultrapure water
- Deionized water
- Waste water
- Highly saline solution, sea water
- Low-ion strength
Accessories & Care
- Immovable glass sleeve junction
- LoT membrane glass
- InLab Pure: refillable FRISCOLYT-B™ (51343185) - keep probe filled with electrolyte for accurate results
- InLab Pure Pro-ISM: refillable 3M KCl (51350072) - keep probe filled with electrolyte for accurate results
- Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
- Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)
Recommended Storage
InLab storage solution

Specifications
Specifications
Connector Types | S7 |
Junction | Immovable Glass Sleeve |
Material | Glass |
pH Range | 1 to 11 |
Type | Combination Electrode |
For Use With (Application) | Measuring samples at low temperatures or with low ion concentrations |
For Use With (Equipment) | S7 to BNC (30281915) |
Internal Reference | ARGENTHAL with Ag+ trap |
Membrane | LoT Glass |
Model | InLab™ Pure |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction