Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

METTLER TOLEDO™ InLab™ Solids pH Electrodes

Catalog No. 01914337 Shop All METTLER TOLEDO Products
Click to view available options
Connector Types:
BNC IP67
Multi-Pin
S7

A low maintenance glass pH electrode with an integrated temperature probe featuring a clog-free, open junction that eliminates the risk of protein fouling. Its spear shaped tip is designed for piercing solid and semi-solid food samples (IP67 rated).

  • For use with METTLER TOLEDO SevenExcellence, SevenCompact and portable meters as well as other brands. Connection cables may be required.
  • Intelligent Sensor Management (ISM) electrodes capture and save calibration data, which is then automatically recognized by the meter once connected.
  • Capture electrode calibration and results with LabX software, designed for advanced data management and 21CFR part 11 regulatory compliance.
  • Compatible with InMotion Autosampler to automate measurement and cleaning of up to 303 samples.

Featured Applications

  • Meats
  • Cheese
  • Seafood
  • Dough
  • Milk and cream
  • Butter, yogurt, ice cream
  • Fruits and vegetables
  • Ready-made meals
  • Solid and semi-solid food samples

Accessories & Care

  • Open junction and non-refillable - no additional electrolyte required
  • Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
  • Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)
TRUSTED_SUSTAINABILITY

Specifications

Description pH Electrode InLab Solids Go-ISM
Connector Types BNC IP67
Junction Open
Material Glass
pH Range 1 to 11
Temperature (Metric) Operating 0°C to 80°C
Type Combination Electrode, Integrated Temperature Probe, Intelligent Sensor Management (ISM) Technology
For Use With (Equipment) Fixed Cable to sensor
Internal Reference ARGENTHAL
Membrane LoT Glass
Model InLab™ Solids Go-ISM
Length (Metric) Cable 1.80 m
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.