Learn More
Restek™ Inlet Adapter Kit for Dual Column Installation
SDP
Catalog No. 06611274 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
For Use With (Equipment):
Agilent Capillary Injectors
Catalog No. For Use With (Equipment)
06-611-274 Agilent Capillary Injectors
1 options
Catalog No. 06-611-274 Supplier Restek™ Supplier No. 27185
Split/Splitless Fitting (1/16 in.) Accepts Standard, Two-Hole Capillary Ferrules
  • Capillary columns are easier to install due to the nut protruding farther from the insulated injection port chamber
  • Same column insertion depth as the original manufacturer's equipment
  • Includes Adaptor Fitting, Capillary Nut (1/16 in.), Gold-Plated Dual Vespel Ring Inlet Seal (1.2 mm I.D.), Two-Hole Ferrules (0.4 mm/0.5 mm I.D.)
Specifications
Type Inlet Adaptor Kit
Includes Adaptor Fitting, Capillary Nut (1/16 in.), Gold-Plated Dual Vespel Ring Inlet Seal (1.2 mm I.D.), Two-Hole Ferrules (0.4 mm/0.5 mm I.D.)
For Use With (Application) Dual Column Installation
For Use With (Equipment) Agilent Capillary Injectors