Learn More
MilliporeSigma™ Supelco™ Inlet Liner, Direct (SPME) Type, Straight Design (unpacked)
Shop All MilliporeSigma Supelco ProductsDescription
- The prime functions of inlet liner are to limit sample degradation and improve vaporisation along with correct positioning of syringe needle.
- The basic problems related with incorrect choice of liner include the higher potential of peak tailing and mass discrimination.
- The inlet liner volume is recommended to be larger than the amount of sample injected by syringe so as to avoid any reaction with the sample.
- Injector port liner is suitable to make connection between sample introduction and GC column.
- Direct type has narrow bore which sustains a high linear velocity through the injection port to diminish band broadening and transferring all the samples to column, flash vaporization.
Specifications
Specifications
| Quantity | 25/Pk. |
| Description | SPME/Splitless |
| Length (Metric) | 74 mm |
| Diameter (Metric) Inner | 0.75 mm |
| Diameter (Metric) Outer | 6.35 mm |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.