Learn More
Restek™ Inlet Maintenance Kit
SDP
Catalog No. 06611205 Shop All Restek Corporation Products
Encompass
Change view
Click to view available options
For Use With (Equipment):
Agilent 5890/6850/6890/7890 GCs
Catalog No. For Use With (Equipment)
06-611-205 Agilent 5890/6850/6890/7890 GCs
1 options
Catalog No. 06-611-205 Supplier Restek™ Supplier No. 22181
For use with Agilent 5890/6850/6890/7890 GCs
  • Includes a variety of the most common consumable GC supplies and tools
  • All parts meet or exceed performance of instrument manufacturer’s parts
  • Parts list makes reordering easy
Specifications
Type Inlet Maintenance Kit
Includes Graphite Ferrules (0.4/0.5/0.8 mm ID), Viton O-Rings, Capillary Nuts, Inlet Seals, Reducing Nut, Scoring Wafer, Thermolite Plus Septa with CenterGuide (11 mm), Topaz Single Taper Liner with Wool (4 mm ID), Topaz Straight Liner with Wool (4 mm ID), Capillary Column Caps, Wrench (1/4 x 5/16 in.), Septum Puller, Installation Gauge, Wire Cleaning Brush, Jet Reamers/Ferrule Removers, Inlet Liner Removal Tool, Septa Nut Removal Tool
Quantity 1 Kit
For Use With (Equipment) Agilent 5890/6850/6890/7890 GCs