Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
INMT Antibody (31G7), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32619750UL
Description
INMT Monoclonal antibody specifically detects INMT in Human samples. It is validated for Flow Cytometry, ELISA, ImmunohistochemistrySpecifications
INMT | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, ELISA, Immunohistochemistry | |
31G7 | |
Flow Cytometry 1:50-1:200, ELISA, Immunohistochemistry 1:50-1:200 | |
Amine N-methyltransferase, Aromatic alkylamine N-methyltransferase, Arylamine N-methyltransferase, EC 2.1.1, EC 2.1.1.49, Indolamine N-methyltransferase, indolethylamine N-methyltransferase, MGC125940, MGC125941, nicotine N-methyltransferase | |
A synthesized peptide derived from Human INMT [UniProt O95050] | |
50 μL | |
Amino Acids Drugs and other small molecules, Endocrinology, Signal Transduction | |
11185 | |
Store at -20 to -70C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction