Learn More
Invitrogen™ INMT Polyclonal Antibody

Description
Immunogen sequence: RVLKCDVHLGN PLAPAVLPLA DCVLTLLAME CACCSLDAYR AALCNLASLL Highest antigen sequence identity to the following orthologs - mouse 67%, rat 67%.
Specifications
Specifications
| Antigen | INMT |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | INMT |
| Gene Accession No. | O95050 |
| Gene Alias | Amine N-methyltransferase; aromatic alkylamine N-methyltransferase; arylamine N-methyltransferase; indolamine N-methyltransferase; Indolethylamine N-methyltransferase; Inmt; nicotine N-methyltransferase; Temt; Thioether S-methyltransferase |
| Gene Symbols | INMT |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.