Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ INMT Recombinant Rabbit Monoclonal Antibody (31G7)
GREENER_CHOICE

Catalog No. FEMA550132
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550132 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550132 Supplier Invitrogen™ Supplier No. MA550132
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

INMT Recombinant Monoclonal Antibody for IHC (P), Flow, ELISA

N-methylation of endogenous and xenobiotic compounds is a major method by which they are degraded. This gene encodes an enzyme that N-methylates indoles such as tryptamine.
TRUSTED_SUSTAINABILITY

Specifications

Antigen INMT
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin)
Classification Recombinant Monoclonal
Clone 31G7
Concentration 1.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene INMT
Gene Accession No. O95050
Gene Alias Amine N-methyltransferase; aromatic alkylamine N-methyltransferase; arylamine N-methyltransferase; indolamine N-methyltransferase; Indolethylamine N-methyltransferase; Inmt; nicotine N-methyltransferase; Temt; Thioether S-methyltransferase
Gene Symbols INMT
Host Species Rabbit
Immunogen A synthesized peptide derived from Human INMT.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11185
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.