Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ InnovaCoat (R) GOLD - Carboxyl 40OD 40nm

Click to view available options
Quantity:
200 μg
40 μg
Specifications
Specifications
Quantity | 40 μg |
Content And Storage | Store at 4°C. Do not freeze. |
Target | InnovaCoat GOLD - Carboxyl 40OD 40nm |
Product Type | Antibody Labeling Kit |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction