Learn More
Novus Biologicals™ InnovaCoat (R) Gold - 20nm Midi Maleimide Antibody Labeling Kit
Shop All R&D Systems Products

Description
- Allows thiolated molecules to be covalently attached to nanoparticles
- Shields biomolecule from metal surface thus eliminating metal-antibody interactions
- Reduces risk of sub-optimal antibody performance
Applications: Electron microscopy visualization
Storage: Store at 4°C
Note: Kits are guaranteed for 6 months from date of receipt
Specifications
Specifications
| Quantity | 50 μg |
| Product Type | 20nm Mini Maleimide Antibody Labeling Kit |
| Sufficient For | 3 Reactions |
| Content And Storage | -20°C |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.