Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ InnovaCoat (R) GOLD 40nm Mini Antibody Labeling Kit
SDP

Catalog No. 2300005 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 Tests
3 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-000-05 3 Tests
23-000-10 10 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 23-000-05 Supplier Novus Biologicals™ Supplier No. 2300005

Antibody labeling kit

InnovaCoat nanoparticles in this kit are supplied as a freeze dried mixture. The conjugation reaction is initiated simply by adding a solution of the antibody, which becomes attached (via lysine residues) to the InnovaCoat surface. InnovaCoat GOLD can however be provided in a variety of formats (e.g. 10 OD liquid or as lyophilized nanoparticles) formulated to allow a one-step reaction with antibodies for bulk or custom orders. The kits are designed for research use and for the development of diagnostic products, specifically for the screening of potential antibodies or small scale proof-of-principle studies. The gold nanoparticles are also designed to be highly effective in lateral flow immunoassays.

Order Info

Quantity

Specifications

Quantity 3 Tests
Content And Storage Store at 4°C. Do not freeze.
Kit Contents 3 Vials of InnovaCoat GOLD, 1 Vial of Reaction Buffer
Product Type InnovaCoat GOLD 40nm Mini Antibody Labeling Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.