Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ InnovaCoat (R) Gold 40nm Streptavidin
SDP

Catalog No. 2500200 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
200 μL
1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-002-00 200 μL
25-010-00 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
For Research Use Only
Catalog No. 25-002-00 Supplier Novus Biologicals™ Supplier No. 2500200
For Research Use Only

Gold nanoparticle with a unique protective surface coat. Covalently attached streptavidin ligand cannot desorb.

  • Can withstand the most extreme conditions (2.5M NaH at 70°C for ≈2 hours)
  • Streptavidin is attached to InnovaCoat™ GOLD via an extended linker to facilitate high-affinity molecular interactions with avidin and streptavidin

Specifications

Content And Storage Store at 4°C. Do not freeze.
Quantity 200 μL
Product Type Antibody Labeling Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.